CAS RN 3092-17-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Midodrine hydrochloride, MilliporeSigma™ Supelco™
Midodrine is a prodrug and a long acting α-adrenergic stimulating agent, and forms an active metabolite, desglymidodrine, which causes an elevation of the systemic blood pressure with the reduction in heart rate.