Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Midodrine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111019176 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9176 50 mg
11-101-8235 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9176 Supplier MilliporeSigma Supelco Supplier No. M827750MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Midodrine is a prodrug and a long acting α-adrenergic stimulating agent, and forms an active metabolite, desglymidodrine, which causes an elevation of the systemic blood pressure with the reduction in heart rate.

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 3092-17-9
Physical Form Neat
Linear Formula C12H18N2O4 · HCl
MDL Number MFCD00079455
Synonym (+/-)-1-(2,5-Dimethoxyphenyl)-2-glycinamidoethanol
Type Analytical Standard
Industry Type Drug Analysis
Quantity 50 mg
Formula Weight 290.74
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.