accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3095-95-2

IUPAC Name: 2-(diethoxyphosphoryl)acetate
Synonyms: 2-(diethoxyphosphoryl)acetate
Molecular Weight (g/mol): 195.13
Molecular Formula: C6H12O5P
InChi Key: DVQMPWOLBFKUMM-UHFFFAOYSA-M
SMILES: CCOP(=O)(CC([O-])=O)OCC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)