Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylphosphonoacetic Acid

Catalog No. 2681275ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-812-75ML 5 mL
26-812-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 26-812-75ML Supplier Sigma Aldrich Supplier No. 2681275ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to make phosphonoester intermediates for intramolecular HEW reactions.

Specifications

CAS 3095-95-2
Density 1.220 g/mL at 25°C
Boiling Point 150°C (0.05 mmHg, lit.)
Molecular Formula C6H13O5P
Linear Formula (C2H5O)2P(O)CH2CO2H
Refractive Index n20/D 1.445
MDL Number MFCD00192032
Quantity 5 mL
Synonym (Diethoxyphosphinyl)acetic acid; Diethyl carboxymethylphosphonate; NSC 272281
Molecular Weight (g/mol) 196.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.