accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSI

CAS RN 30955-94-3

IUPAC Name: 1-(5-iodothiophen-2-yl)ethan-1-one
Synonyms: 1-(5-iodothiophen-2-yl)ethanone
Molecular Weight (g/mol): 252.07
Molecular Formula: C6H5IOS
InChi Key: XBUCSKKBSJSRLJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(I)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results