accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3096-69-3

IUPAC Name: 4-amino-2,3-dimethylphenol
Synonyms: 4-amino-2,3-dimethylphenol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: UBKPLLYABUUFCE-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(O)=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results