Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,3-xylenol 98.0+%, TCI America™
SDP

Catalog No. A096110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A096110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A096110G Supplier TCI America Supplier No. A096110G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3096-69-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00070554
InChI Key UBKPLLYABUUFCE-UHFFFAOYSA-N
Synonym 4-Amino-2,3-dimethylphenol, 4-Hydroxy-2,3-dimethylaniline
PubChem CID 76542
IUPAC Name 4-amino-2,3-dimethylphenol
SMILES CC1=C(C=CC(=C1C)O)N

Specifications

Melting Point 174°C
Quantity 10 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,3-xylenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.