accessibility menu, dialog, popup

UserName

Viewing profile as user:
KH3CCH2OOSOOO

CAS RN 31098-21-2

IUPAC Name: potassium 3-[(2-methylprop-2-enoyl)oxy]propane-1-sulfonate
Synonyms: potassium 3-[(2-methylprop-2-enoyl)oxy]propane-1-sulfonate
Molecular Weight (g/mol): 246.32
Molecular Formula: C7H11KO5S
InChi Key: PNOXUQIZPBURMT-UHFFFAOYSA-M
SMILES: [K+].CC(=C)C(=O)OCCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)