accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 311-28-4

IUPAC Name: tetrabutylazanium iodide
Synonyms: tetrabutylammonium ion iodide
Molecular Weight (g/mol): 369.38
Molecular Formula: C16H36IN
InChi Key: DPKBAXPHAYBPRL-UHFFFAOYSA-M
SMILES: [I-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (12)
Quantity 
  • (1)
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (7)
Physical Form 
  • (7)
  • (1)
  • (2)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (1)
  • (6)
  • (3)
  • (7)
  • (1)
  • (6)
  • (12)
  • (1)
  • (5)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (7)
  • (7)
  • (1)
  • (2)
  • (2)
  • (1)