Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Tetrabutylammonium iodide,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013895 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3895 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3895 Supplier MilliporeSigma Supelco Supplier No. 8690310GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00011636
Synonym TBAI

Specifications

Melting Point 141°C to 143°C; 144°C to 146°C
CAS 311-28-4
Physical Form Crystals
Quantity 10 g
Linear Formula (CH3 CH2 CH2 CH2)4 N(I)
Molecular Weight (g/mol) 369.37
Formula Weight 369.37
Percent Purity ≥99.0% (AT); ≥99.0%
Grade Ion Pair Chromatography
Product Line LiChropur
Chemical Name or Material Tetrabutylammonium Iodide
Show More Show Less
RTECSNumber BS5450000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.