CAS RN 31105-90-5
CAS RN 31105-90-5
IUPAC Name:
(2S)-2-amino-3-(3,4-difluorophenyl)propanoic acid
Synonyms:
(2S)-2-amino-3-(3,4-difluorophenyl)propanoic acid
Molecular Weight (g/mol):
201.17
Molecular Formula:
C9H9F2NO2
InChi Key:
PRAWYXDDKCVZTL-QMMMGPOBSA-N
SMILES:
N[C@@H](CC1=CC=C(F)C(F)=C1)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech L-Phe 3.4-diF , Advanced ChemTech
CAS: 31105-90-5 Molecular Formula: C9H9F2NO2 Molecular Weight (g/mol): 201.17 InChI Key: PRAWYXDDKCVZTL-QMMMGPOBSA-N IUPAC Name: (2S)-2-amino-3-(3,4-difluorophenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(F)C(F)=C1)C(O)=O