accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31112-62-6

IUPAC Name: 3-acetamido-2,4,6-triiodo-5-(N-methylacetamido)-N-[(2S,5S)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]benzamide
Synonyms: metrizamide
Molecular Weight (g/mol): 789.1
Molecular Formula: C18H22I3N3O8
InChi Key: BAQCROVBDNBEEB-OPRFMTNSSA-N
SMILES: CN(C(C)=O)C1=C(I)C(NC(C)=O)=C(I)C(C(=O)NC2[C@@H](O)OC(CO)[C@@H](O)C2O)=C1I
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)