Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metrizamide p.a., Thermo Scientific™

Catalog No. AC229430250
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC229430250 25 g
AC229430010 1 g
AC229430100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC229430250 Supplier Thermo Scientific Chemicals Supplier No. 229430250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31112-62-6
Molecular Formula C18H22I3N3O8
Molecular Weight (g/mol) 789.1
InChI Key BAQCROVBDNBEEB-UBLAARQISA-N
Synonym metrizamide, amipaque, telebrix 300, 2-3-acetylamino-5-acetylmethylamino-2,4,6-triiodobenzoyl amino-2-deoxy-d-glucose
PubChem CID 45039930
IUPAC Name 3-acetamido-5-[acetyl(methyl)amino]-2,4,6-triiodo-N-[(3S,5S)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]benzamide
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)NC2C(C(C(OC2O)CO)O)O)I)N(C)C(=O)C)I

Specifications

Absorbance 0.07 max. (62% in water) at 420nm
Color White
Infrared Spectrum Authentic
Quantity 25 g
Merck Index 15, 6234
Specific Rotation +17° to +19° (20°C, 589nm) (c=5, water)
Formula Weight 789.09
Percent Purity ≥99% (on dry substance)
Physical Form Fine Crystalline Powder
Chemical Name or Material Metrizamide, p.a.
EINECSNumber 250-475-5
RTECSNumber LZ6650000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.