accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3112-88-7

IUPAC Name: [(benzenesulfonyl)methyl]benzene
Synonyms: benzyl phenyl sulfone
Molecular Weight (g/mol): 232.3
Molecular Formula: C13H12O2S
InChi Key: FABCMLOTUSCWOR-UHFFFAOYSA-N
SMILES: O=S(=O)(CC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results