accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 312-21-0

IUPAC Name: 4-trifluoromethanesulfonylbenzonitrile
Synonyms: 4-trifluoromethanesulfonylbenzonitrile
Molecular Weight (g/mol): 235.18
Molecular Formula: C8H4F3NO2S
InChi Key: UHCGSMMNICMDIX-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results