accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOOHN

CAS RN 31249-95-3

IUPAC Name: 1,4,10-trioxa-7,13-diazacyclopentadecane
Synonyms: 1,4,10-trioxa-7,13-diazacyclopentadecane
Molecular Weight (g/mol): 218.3
Molecular Formula: C10H22N2O3
InChi Key: STHIZMRUXPMSCW-UHFFFAOYSA-N
SMILES: C1COCCNCCOCCOCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results