Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,10-Diaza-15-crown 5-Ether 97.0+%, TCI America™
SDP

Catalog No. D27441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27441G Supplier TCI America Supplier No. D27441G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31249-95-3
Molecular Formula C10H22N2O3
Molecular Weight (g/mol) 218.297
MDL Number MFCD00005109
InChI Key STHIZMRUXPMSCW-UHFFFAOYSA-N
Synonym 1,4,10-trioxa-7,13-diaza-cyclopentadecane, cryptand 21, kryptofix 21, 4,10-diaza-15-crown 5-ether, 1,7,13-trioxa-4,10-diazacyclopentadecane, 4,7,13-trioxa-1,10-diazacyclopentadecane, kryptofix™ 21, acmc-1cre7, 1,7-diaza-15-crown-5, 1,7,10-trioxa-4,13-diazacyclopentadecane
PubChem CID 123435
IUPAC Name 1,4,10-trioxa-7,13-diazacyclopentadecane
SMILES C1COCCNCCOCCOCCN1

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 1 g
Formula Weight 218.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,10-Diaza-15-crown 5-Ether
RTECSNumber YJ9520000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.