accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH3COOH3COOH3COHo

CAS RN 312619-49-1

IUPAC Name: holmium(3+) triacetate
Synonyms: holmium(3+) triacetate
Molecular Weight (g/mol): 342.06
Molecular Formula: C6H9HoO6
InChi Key: HWQRVFRWLQGBFV-UHFFFAOYSA-K
SMILES: [Ho+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)