accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 312619-56-0

IUPAC Name: 3-methylbenzene-1-sulfonic acid hydrate
Synonyms: 3-methylbenzenesulfonic acid hydrate
Molecular Weight (g/mol): 190.21
Molecular Formula: C7H10O4S
InChi Key: GHLYGGIKYFLYTH-UHFFFAOYSA-N
SMILES: O.CC1=CC(=CC=C1)S(O)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results