accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNOOOH(E/Z)

CAS RN 312694-02-3

IUPAC Name: 6-{2-propyl-4-[2-(pyridin-4-yl)diazen-1-yl]phenoxy}hexanoic acid
Synonyms: 6-{2-propyl-4-[2-(pyridin-4-yl)diazen-1-yl]phenoxy}hexanoic acid
Molecular Weight (g/mol): 355.44
Molecular Formula: C20H25N3O3
InChi Key: RKSDFQDWVOJLAU-UHFFFAOYSA-N
SMILES: CCCC1=C(OCCCCCC(O)=O)C=CC(=C1)N=NC1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results