accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOH2N

CAS RN 3129-91-7

IUPAC Name: N-cyclohexylcyclohexanaminium nitrite
Synonyms: dicyclohexylammonium nitrite
Molecular Weight (g/mol): 228.34
Molecular Formula: C12H24N2O2
InChi Key: ZFAKTZXUUNBLEB-UHFFFAOYSA-N
SMILES: [O-]N=O.C1CCC(CC1)[NH2+]C1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)