Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicyclohexylamine Nitrite 97.0+%, TCI America™
SDP

Catalog No. D1392500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1392500G 500 g
D139225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1392500G Supplier TCI America Supplier No. D1392500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3129-91-7
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD00013128
InChI Key ZFAKTZXUUNBLEB-UHFFFAOYSA-N
PubChem CID 241753
IUPAC Name N-cyclohexylcyclohexanaminium nitrite
SMILES [O-]N=O.C1CCC(CC1)[NH2+]C1CCCCC1

Specifications

Melting Point 183°C
Color White
Quantity 500 g
UN Number 2687
Formula Weight 228.34
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylamine Nitrite
RTECSNumber HY4200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.