accessibility menu, dialog, popup

UserName

Viewing profile as user:
BClFFFHOOH

CAS RN 313545-41-4

IUPAC Name: [4-chloro-2-(trifluoromethyl)phenyl]boronic acid
Synonyms: 4-chloro-2-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 224.37
Molecular Formula: C7H5BClF3O2
InChi Key: YAPBOBGBYQQYHX-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=C(Cl)C=C1)C(F)(F)F
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)