Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C26015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2601-5G 5 g
C26011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2601-5G Supplier TCI America Supplier No. C26015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 313545-41-4
Melting Point 182°C
Color White-Yellow
Molecular Formula C7H5BClF3O2
Quantity 5 g
MDL Number MFCD04973086
Synonym 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethylphenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl-phenylboronic acid
InChI Key YAPBOBGBYQQYHX-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
IUPAC Name [4-chloro-2-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 224.37
PubChem CID 2763244
Formula Weight 224.37
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.