accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 313545-44-7

IUPAC Name: (2-chloro-4-ethoxyphenyl)boronic acid
Synonyms: 2-chloro-4-ethoxyphenylboronic acid
Molecular Weight (g/mol): 200.43
Molecular Formula: C8H10BClO3
InChi Key: GMFQJZUNQAJZDH-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(B(O)O)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results