accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NNBr

CAS RN 313735-62-5

IUPAC Name: 4-bromo-1-(propan-2-yl)-1H-pyrazole
Synonyms: 4-bromo-1-isopropylpyrazole
Molecular Weight (g/mol): 189.06
Molecular Formula: C6H9BrN2
InChi Key: HYWPFIXULAMLRZ-UHFFFAOYSA-N
SMILES: CC(C)N1C=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results