Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-1-isopropylpyrazole 98.0+%, TCI America™
SDP

Catalog No. B51801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B51801G Supplier TCI America Supplier No. B51801G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 313735-62-5
Molecular Formula C6H9BrN2
Molecular Weight (g/mol) 189.056
MDL Number MFCD09801021
InChI Key HYWPFIXULAMLRZ-UHFFFAOYSA-N
PubChem CID 22329200
IUPAC Name 4-bromo-1-propan-2-ylpyrazole
SMILES CC(C)N1C=C(C=N1)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 189.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-isopropylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.