accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNS

CAS RN 3138-80-5

IUPAC Name: 1H,3H-thieno[3,4-b]quinoxaline
Synonyms: 1H,3H-thieno[3,4-b]quinoxaline
Molecular Weight (g/mol): 188.25
Molecular Formula: C10H8N2S
InChi Key: OSKBIJDQSJQRQK-UHFFFAOYSA-N
SMILES: C1SCC2=C1N=C1C=CC=CC1=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results