Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dihydrothieno[3,4-b]quinoxaline, 97%, Thermo Scientific™

Catalog No. AAH5890509
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5890509 10 g
AAH5890504 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5890509 Supplier Thermo Scientific Chemicals Supplier No. H5890509

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3138-80-5
Molecular Formula C10H8N2S
Molecular Weight (g/mol) 188.248
MDL Number MFCD22683025
InChI Key OSKBIJDQSJQRQK-UHFFFAOYSA-N
Synonym 1,3-dihydrothieno 3,4-b quinoxaline, thieno 3,4-b quinoxaline, 1,3-dihydro, 1h,3h-thieno 3,4-b quinoxaline
PubChem CID 11095365
IUPAC Name 1,3-dihydrothieno[3,4-b]quinoxaline
SMILES C1C2=NC3=CC=CC=C3N=C2CS1

Specifications

Quantity 10 g
Formula Weight 188.25
Percent Purity 97%
Chemical Name or Material 1,3-Dihydrothieno[3,4-b]quinoxaline
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.