accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3138-90-7

IUPAC Name: 1-benzyl-3-methylpiperazine
Synonyms: 1-benzyl-3-methylpiperazine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: QOFUDSPYJDXBOF-UHFFFAOYNA-N
SMILES: CC1CN(CC2=CC=CC=C2)CCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results