accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOIIOOOH

CAS RN 31395-16-1

IUPAC Name: 3',6'-dihydroxy-4,5-diiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 3',6'-dihydroxy-4,5-diiodospiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 584.1
Molecular Formula: C20H10I2O5
InChi Key: RUEQYKGDINYYHI-UHFFFAOYSA-N
SMILES: OC1=CC2=C(C=C1)C1(OC(=O)C3=C1C=CC(I)=C3I)C1=C(O2)C=C(O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results