CAS RN 3140-73-6
IUPAC Name:
2-chloro-4,6-dimethoxy-1,3,5-triazine
Synonyms:
2-chloro-4,6-dimethoxy-1,3,5-triazine
Molecular Weight (g/mol):
175.57
Molecular Formula:
C5H6ClN3O2
InChi Key:
GPIQOFWTZXXOOV-UHFFFAOYSA-N
SMILES:
COC1=NC(OC)=NC(Cl)=N1
1
–
4
of
4
results
1
Promotions Available
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%
2
Promotions Available
2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%