accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNHNNO

CAS RN 31431-39-7

IUPAC Name: methyl N-(6-benzoyl-1H-1,3-benzodiazol-2-yl)carbamate
Synonyms: mebendazole verpanyl mebutar vermicidin telmin besantin pantelmin vermox (TN) vermirax noverme
Molecular Weight (g/mol): 295.3
Molecular Formula: C16H13N3O3
InChi Key: OPXLLQIJSORQAM-UHFFFAOYSA-N
SMILES: COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=CC=C1
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)