Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mebendazole, 98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012193 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2193 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2193 Supplier MilliporeSigma Supelco Supplier No. M252325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mebendazole is a benzimidazole carbamate derivative, which is an anthelmintic agent.

TRUSTED_SUSTAINABILITY

Specifications

CAS 31431-39-7
Physical Form Neat
Linear Formula C16H13N3O3
MDL Number MFCD00057872
Synonym 5-Benzoyl-2-benzimidazolecarbamic acid methyl ester; Mebendazol; Methyl N-(5-benzoyl-1 H-benzimidazol-2-yl)carbamate
Type Analytical Standard
Quantity 25 g
Formula Weight 295.29
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.