accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNH2OO

CAS RN 3144-09-0

IUPAC Name: methanesulfonamide
Synonyms: methanesulfonamide
Molecular Weight (g/mol): 95.12
Molecular Formula: CH5NO2S
InChi Key: HNQIVZYLYMDVSB-UHFFFAOYSA-N
SMILES: CS(N)(=O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)