accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31457-16-6

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-8-carboxylic acid
Synonyms: 3,4-dihydro-2H-1-benzopyran-8-carboxylic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: LOFOWPRKKPHPDW-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2OCCCC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results