accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31571-14-9

IUPAC Name: 3-(hydroxyimino)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: 3-(hydroxyimino)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: YRNPDSREMSMKIY-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=O)C2=NO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results