accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNOO

CAS RN 31594-45-3

IUPAC Name: 2-ethoxy-5-nitropyridine
Synonyms: 2-ethoxy-5-nitropyridine
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: LYDVDLYMQVSLQD-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=N1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results