Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxy-5-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. E05365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05365G 5 g
E053625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05365G Supplier TCI America Supplier No. E05365G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31594-45-3
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD01646181
InChI Key LYDVDLYMQVSLQD-UHFFFAOYSA-N
PubChem CID 295864
IUPAC Name 2-ethoxy-5-nitropyridine
SMILES CCOC1=CC=C(C=N1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxy-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.