CAS RN 316130-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ AMPK Activator VI, RSVA314
The AMPK Activator VI, RSVA314 modulates the biological activity of AMPK. This small molecule/inhibitor is primarily used for Biochemicals applications.