accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 316808-41-0

IUPAC Name: 1,8-bis(diphenylphosphoroso)naphthalene
Synonyms: 1,8-bis(diphenylphosphoroso)naphthalene
Molecular Weight (g/mol): 528.53
Molecular Formula: C34H26O2P2
InChi Key: XNKIEKXYLNHGBX-UHFFFAOYSA-N
SMILES: O=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC2=CC=CC(=C12)P(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results