Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Bis(diphenylphosphinyl)naphthalene 98.0+%, TCI America™
SDP

Catalog No. B52805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52805G 5 g
B52801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52805G Supplier TCI America Supplier No. B52805G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 316808-41-0
Molecular Formula C34H26O2P2
Molecular Weight (g/mol) 528.528
InChI Key XNKIEKXYLNHGBX-UHFFFAOYSA-N
Synonym Naphthalene-1,8-diylbis(diphenylphosphine Oxide)
PubChem CID 58555919
IUPAC Name 1,8-bis(diphenylphosphoryl)naphthalene
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)C3=CC=CC4=C3C(=CC=C4)P(=O)(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Color Yellow
Quantity 5 g
Formula Weight 528.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(diphenylphosphinyl)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.