accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3COOH

CAS RN 31728-75-3

IUPAC Name: 1,5-dimethyl-1H-pyrazole-4-carboxylic acid
Synonyms: 1,5-dimethylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: SKTIXLZNJZTGRT-UHFFFAOYSA-N
SMILES: CN1N=CC(C(O)=O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results