Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Do not store together with acids. Incompatible with oxidizing agents, bases, acids.
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Do not store together with acids. Incompatible with oxidizing agents, bases, acids.
Specifications
Specifications
| CAS | 31728-75-3 |
| Molecular Formula | C6H8N2O2 |
| Quantity | 5g |
| MDL Number | MFCD02090876 |
| Synonym | 1,5-dimethyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-4-carboxylicacid, 1,5-dimethyl |
| InChI Key | SKTIXLZNJZTGRT-UHFFFAOYSA-N |
| SMILES | CN1N=CC(C(O)=O)=C1C |
| IUPAC Name | 1,5-dimethylpyrazole-4-carboxylic acid |
| Molecular Weight (g/mol) | 140.14 |
| PubChem CID | 1273303 |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?