accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 317385-62-9

H3COOS

CAS RN 317385-62-9

IUPAC Name: butyl thiophene-3-carboxylate
Synonyms: butyl thiophene-3-carboxylate
Molecular Weight (g/mol): 184.25
Molecular Formula: C9H12O2S
InChi Key: PNELMESXUGBLMJ-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results