Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl 3-Thiophenecarboxylate, TCI America™
SDP

Catalog No. T19641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19641G Supplier TCI America Supplier No. T19641G

Chemical Identifiers

CAS 317385-62-9
Molecular Formula C9H12O2S
Molecular Weight (g/mol) 184.253
MDL Number MFCD04038430
InChI Key PNELMESXUGBLMJ-UHFFFAOYSA-N
Synonym 3-Thiophenecarboxylic Acid Butyl Ester
PubChem CID 20277519
IUPAC Name butyl thiophene-3-carboxylate
SMILES CCCCOC(=O)C1=CSC=C1

Specifications

Quantity 1 g
Formula Weight 184.25
Chemical Name or Material Butyl 3-Thiophenecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.