accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 317802-08-7

IUPAC Name: 2-[7-(1,3,2-dioxaborinan-2-yl)-9,9-dioctyl-9H-fluoren-2-yl]-1,3,2-dioxaborinane
Synonyms: 2-[7-(1,3,2-dioxaborinan-2-yl)-9,9-dioctylfluoren-2-yl]-1,3,2-dioxaborinane
Molecular Weight (g/mol): 558.42
Molecular Formula: C35H52B2O4
InChi Key: KAYXDWIILRESPY-UHFFFAOYSA-N
SMILES: CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B2OCCCO2)C2=C1C=C(C=C2)B1OCCCO1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results