Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 9,9-Dioctylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester

Catalog No. 5693565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-935-65G 5 g
56-935-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-935-65G Supplier Sigma Aldrich Supplier No. 5693565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

9,9-Dioctylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester

Specifications

CAS 317802-08-7
Melting Point 126°C to 130°C
Molecular Formula C35H52B2O4
Linear Formula C3H6O2BC6H3C(C8H17)2C6H3BO2C3H6
Quantity 5 g
MDL Number MFCD03701607
Synonym 9,9-Dioctylfluorene-2,7-bis(trimethylborate)
Molecular Weight (g/mol) 558.41
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.