accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 317830-83-4

IUPAC Name: (1-benzofuran-3-yl)boronic acid
Synonyms: 1-benzofuran-3-ylboronic acid
Molecular Weight (g/mol): 161.95
Molecular Formula: C8H7BO3
InChi Key: DFUGYZQSDFQVPU-UHFFFAOYSA-N
SMILES: OB(O)C1=COC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (1)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)