Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzofuran-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B45225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4522-5G 5 g
B45221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4522-5G Supplier TCI America Supplier No. B45225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 317830-83-4
Molecular Formula C8H7BO3
MDL Number MFCD06801687
InChI Key DFUGYZQSDFQVPU-UHFFFAOYSA-N
PubChem CID 23438904

Specifications

Quantity 5 g
Synonym benzofuran-3-ylboronic acid, benzofuran-3-boronic acid, 1-benzofuran-3-yl boronic acid, boronic acid, 3-benzofuranyl, acmc-1 csxn
SMILES B(C1=COC2=CC=CC=C12)(O)O
IUPAC Name 1-benzofuran-3-ylboronic acid
Molecular Weight (g/mol) 161.951
Formula Weight 161.95
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.